missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UMPS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14830-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Involved in signal transduction through the Wnt pathway.Spécification
UMPS | |
Polyclonal | |
Unconjugated | |
UMPS | |
OMPdecase, OPRT, OPRTase, UMPS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22247, 288051, 7372 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P11172, P13439 | |
Umps | |
UMPS Fusion Protein Ag6620 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |