missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UQCC2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25781-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes a nucleoid protein localized to the mitochondria inner membrane. The encoded protein affects regulation of insulin secretion, mitochondrial ATP production, and myogenesis through modulation of mitochondrial respiratory chain activity.Spécification
UQCC2 | |
Polyclonal | |
Unconjugated | |
UQCC2 | |
C6orf125, uqcc2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84300 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BRT2 | |
UQCC2 | |
UQCC2 Fusion Protein Ag22710 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |