missing translation for 'onlineSavingsMsg'
Learn More

Urocortin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 19731-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16829214

  • 325.38€ / 150µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Urocortin (UCN), a 40 amino acids neuropeptide widely distributed in the central nervous system (CNS), belongs to a family of structurally related peptides including corticotrophin releasing factor (CRF), sauvagine and urotensin I. Urocortin is derived from a larger precursor protein (pro-urocortin, also named corticotensin). Urocortin has biological actions similar to CRF such as stress-related endocrine, autonomic and behavioral responses in mammalian species. It is a potent stimulator of adrenocorticotropic hormone (ACTH) secretion by corticotroph cells from the anterior pituitary in vitro and in vivo.
Spécification

Spécification

Urocortin
Polyclonal
Unconjugated
UCN
UCN, UI, UROC, urocortin
Rabbit
Antigen Affinity Chromatography
RUO
22226, 29151, 7349
-20°C
Liquid
Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P55089, P55090, P81615
Ucn
Peptide
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documenti
Product Certifications
Offerte speciali

Offerte speciali