missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UVRAG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
121.38€ - 325.38€
Spécification
Antigène | UVRAG |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16839114
|
Proteintech
19571-1-AP-20UL |
20 μL |
Cet article est actuellement indisponible ou discontinu.
Veuillez consulter la page produit pour de possible alternative dans l’onglet |
||||||
16829114
|
Proteintech
19571-1-AP-150UL |
150 μL |
Cet article est actuellement indisponible ou discontinu.
Veuillez consulter la page produit pour de possible alternative dans l’onglet |
||||||
Description
UVRAG is predicted to contain a coiled-coil structure and is likely to be metabolically unstable based on the occurrence of a strong PEST domain. It interacts with Beclin 1, leading to activation of autophagy and thereof inhibition of tumorigenicity of human colon cancer cells. The gene for UVRAG protein is localized on human chromosome 11q13.5 and complements the ultraviolet sensitivity of xeroderma pigmentosum group C cells. The gene has been extremely well conserved during evolution, implying an important role for this gene product in cell metabolism. Chromosomal aberrations involving this gene are associated with left-right axis malformation, and mutations in this gene have been associated with colon cancer. The protein is highly expressed in human brain, lung, kidney and liver.Spécification
UVRAG | |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q8K245, Q9P2Y5 | |
308846, 7405, 78610 | |
UVRAG Fusion Protein Ag13765 | |
Primary | |
-20°C | |
UVRAG |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DHTX, p63, UVRAG | |
Uvrag | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |