missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VIP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA576117
Informations supplémentaires : Poids : 0.01000kg
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Vasoactive intestinal peptide (VIP) is a 28-amino-acid peptide belonging to the secretin-glucagon family of peptide hormones. Its sequence is identical in man, ox, pig and rat. VIP is widely distributed in the central and peripheral nervous systems, acting as a neurotransmitter or neuromodulator. Peripherally it is involved in vaso- and bronchodilatation, smooth muscle relaxation and stimulation of secretion. Ectopic endocrine tumors secreting VIP (VIPomas) give rise to raised plasma levels of VIP and are associated with watery diarrhea, hypokalemia and achlorhydria syndrome.Spécification
VIP | |
Polyclonal | |
Unconjugated | |
VIP | |
VIP peptides; PHM27; | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
117064, 22353, 7432 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P01282, P01283, P32648 | |
VIP | |
Recombit protein of human VIP. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |