Learn More
VIP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA5120606
Informations supplémentaires : Poids : 0.01000kg
Description
Positive test controls include: BxPC-3, U-87MG, SH-SY5Y, Mouse spleen, Mouse brain, Rat brain. The target is usually found in the following locations: Secreted. Immunogen sequence: MDTRNKAQLL VLLTLLSVLF SQTSAWPLYR APSALRLGDR IPFEGANEPD QVSLKEDIDM LQNALAENDT PYYDVSRNAR HADGVFTSDF SKLLGQLSAK KYLESLMGKR VSNISEDPVP VKRHSDAVFT DNYTRLRKQM AVKKYLNSIL NGKRSSEGES PDFPEELEK
VIP encodes a protein that belongs to the glucagon family. It stimulates myocardial contractility, causes vasodilation, increases glycogenolysis, lowers arterial blood pressure and relaxes the smooth muscle of trachea, stomach and gall bladder. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.Spécification
VIP | |
Polyclonal | |
Unconjugated | |
VIP | |
Intestinal peptide PHI-27; Intestinal peptide PHI-42; Intestinal peptide PHM-27; Intestinal peptide PHV-42; MGC13587; PACAP-R-2; Peptide histidine isoleucinamide 27; Peptide histidine methioninamide 27; Peptide histidine valine 42; PHI; PHI, peptide histidine isoleucine; PHM27; prepro-VIP; vasoactive intestinal peptide; Vasoactive intestinal polypeptide; VIP; VIP peptides; vip/phi27; VIP-R-1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
117064, 22353, 7432 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.59 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P01282, P01283, P32648 | |
VIP | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-169 of human VIP (NP_9194161) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |