Learn More
VIP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA585617
Informations supplémentaires : Poids : 0.01000kg
Description
Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Pig (100%), Sheep (100%), Rhesus Monkey (100%), Bovine (100%). Positive Control: SK-N-AS. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
VIP encodes a protein that belongs to the glucagon family. It stimulates myocardial contractility, causes vasodilation, increases glycogenolysis, lowers arterial blood pressure and relaxes the smooth muscle of trachea, stomach and gall bladder. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.Spécification
VIP | |
Polyclonal | |
Unconjugated | |
VIP | |
Intestinal peptide PHI-27; Intestinal peptide PHI-42; Intestinal peptide PHM-27; Intestinal peptide PHV-42; MGC13587; PACAP-R-2; Peptide histidine isoleucinamide 27; Peptide histidine methioninamide 27; Peptide histidine valine 42; PHI; PHI, peptide histidine isoleucine; PHM27; prepro-VIP; vasoactive intestinal peptide; Vasoactive intestinal polypeptide; VIP; VIP peptides; vip/phi27; VIP-R-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22353, 7432 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Western Blot | |
0.94 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P01282, P32648 | |
VIP | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human VIP. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |