missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VIP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA586514
Informations supplémentaires : Poids : 0.01000kg
Description
VIP encodes a protein that belongs to the glucagon family. It stimulates myocardial contractility, causes vasodilation, increases glycogenolysis, lowers arterial blood pressure and relaxes the smooth muscle of trachea, stomach and gall bladder. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.Spécification
VIP | |
Polyclonal | |
Unconjugated | |
VIP | |
Intestinal peptide PHI-27, Intestinal peptide PHI-42, Intestinal peptide PHM-27, Intestinal peptide PHV-42, MGC13587, PACAP-R-2, Peptide histidine isoleucinamide 27, Peptide histidine methioninamide 27, Peptide histidine valine 42, PHI, PHI, peptide histidine isoleucine, PHM27, prepro-VIP, vasoactive intestinal peptide, Vasoactive intestinal polypeptide, VIP, VIP peptides, vip/phi27, VIP-R-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7432 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide, pH 7.2 | |
P01282 | |
VIP | |
Synthetic peptide, corresponding to amino acids 91-140 of Human VIP. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |