missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WSTF Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-95129-0.1ml
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
WSTF Polyclonal antibody specifically detects WSTF in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| WSTF | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| bromodomain adjacent to zinc finger domain, 1B, williams-Beuren syndrome chromosomal region 10 protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human WSTF (NP_115784.1). MAPLLGRKPFPLVKPLPGEEPLFTIPHTQEAFRTREEYEARLERYSERIWTCKSTGSSQLTHKEAWEEEQEVAELLKEEFPAWYEKLVLEMVHHNTASLEKLVDTAWLEIMTKYAVGEECDFEVGKEKMLKVKIVKIHPLEKVDEEATEKKSDGACDSPSSDKENSSQIAQDHQKKETVVKEDEGRRESINDRARRSPRKLPTSLKKGERKWAPPKFLPHKYDVKLQNEDKIISNVPADSLIRTERPPNKEIVRYFIRHN | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| 9031 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction