missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WWC2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24750-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of the WW-and-C2-domain-containing family of proteins. Members of this family have two N-terminal WW domains that mediate binding to target proteins harboring L/PPxY motifs, an internal C2 domain for membrane association, and C-terminal alpha protein kinase C binding sites and class III PDZ domain-interaction motifs. Proteins of this family are able to form homo- and heterodimers and to modulate hippo pathway signaling.Spécification
WWC2 | |
Polyclonal | |
Unconjugated | |
WWC2 | |
BH 3 only member B, BOMB, Protein WWC2, WW and C2 domain containing 2, WW domain containing protein 2, WWC2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
52357, 80014 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6AWC2, Q6NXJ0 | |
Wwc2 | |
WWC2 Fusion Protein Ag20479 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |