Learn More
XBP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24168-1-AP-150UL
Additional Details : Poids : 0.00010kg
Description
The XBP1 antibody (24168-1-AP) can detect both isoforms XBP1(U) and XBP1(S).
XBP1 encodes a transcription factor that regulates MHC class II genes by binding to a promoter element referred to as an X box. This gene product is a bZIP protein, which was also identified as a cellular transcription factor that binds to an enhancer in the promoter of the T cell leukemia virus type 1 promoter. It may increase expression of viral proteins by acting as the DNA binding partner of a viral transactivator. It has been found that upon accumulation of unfolded proteins in the endoplasmic reticulum (ER), the mRNA of this gene is processed to an active form by an unconventional splicing mechanism that is mediated by the endonuclease inositol-requiring enzyme 1 (IRE1). The resulting loss of 26 nt from the spliced mRNA causes a frame-shift and an isoform XBP1(S), which is the functionally active transcription factor. The isoform encoded by the unspliced mRNA, XBP1(U), is constitutively expressed, and thought to function as a negative feedback regulator of XBP1(S), which shuts off transcription of target genes during the recovery phase of ER stress. A pseudogene of XBP1 has been identified and localized to chromosome 5.Specifications
XBP1 | |
Polyclonal | |
Unconjugated | |
XBP1 | |
TREB5, X box binding protein 1, XBP 1, XBP1, XBP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22433, 7494 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35426, P17861 | |
XBP1 | |
XBP1 Fusion Protein Ag21454 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |