missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XRN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23108-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody is specific to the XRN1 protein.
SEC24C is a member of the SEC24 subfamily of the SEC23/SEC24 family, which is involved in vesicle trafficking. The encoded protein has similarity to yeast Sec24p component of COPII. COPII is the coat protein complex responsible for vesicle budding from the ER. The product of this gene may play a role in shaping the vesicle, as well as in cargo selection and concentration.Spécification
| XRN1 | |
| Polyclonal | |
| Unconjugated | |
| XRN1 | |
| 5 3 exoribonuclease 1, 5′ 3′ exoribonuclease 1, SEP1, XRN1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 24127, 54464 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P97789, Q8IZH2 | |
| XRN1 | |
| XRN1 Fusion Protein Ag19398 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu