missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF174 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 18157-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
ZNF174, also known as ZSCAN8 or AW-1, is a transcriptional repressor that belongs to the Krueppel C2H2-type zinc-finger protein family. It is expressed in a number of different tissues, including small intestine, prostate, colon, spleen, pancreas, skeletal muscle, brain, heart, kidney and thymus, but it is most predomitly found in adult ovary and testis. ZNF174 specifically acts to repress the promoter activities of PDGF-B and TGF beta1. ZNF174 localizes to the nucleus and contains three C2H2-type zinc fingers at the C-terminus and one SCAN domain near the N-terminus. SCAN domains are found in a number of zinc-finger proteins and are characterized by a conserved region of 84 residues. The SCAN domain seemingly regulates the association of proteins containing SCAN domains into noncovalent complexes and may also function as an underlying mechanism in selective oligomerization of these proteins.Spécification
ZNF174 | |
Polyclonal | |
Unconjugated | |
ZNF174 | |
AW 1, zinc finger protein 174, ZNF174, ZSCAN8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7727 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15697 | |
ZNF174 | |
ZNF174 Fusion Protein Ag12963 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |