missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF311 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23243-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody specifically reacts with ZNF311 protein.
ZNF311 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.Spécification
ZNF311 | |
Polyclonal | |
Unconjugated | |
ZNF311 | |
zf31, ZFP31, zinc finger protein 311, Zinc finger protein zfp 31, ZNF311 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
282890 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5JNZ3 | |
ZNF311 | |
ZNF311 Fusion Protein Ag19624 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |