missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF419 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25724-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
May be involved in transcriptional regulation.Spécification
ZNF419 | |
Polyclonal | |
Unconjugated | |
ZNF419 | |
zinc finger protein 419, ZNF419, ZNF419A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79744 | |
-20°C | |
Liquid |
Western Blot | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96HQ0 | |
ZNF419 | |
ZNF419 Fusion Protein Ag22802 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |