missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZXDA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26261-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes one of two duplicated zinc finger genes on chromosome Xp11. This gene is the telomeric copy; GeneID 158586 ZXDB is the more centromeric copy. The two genes have 98% nucleotide sequence similarity, and the predicted proteins contain 10 tandem zinc finger motifs.Spécification
ZXDA | |
Polyclonal | |
Unconjugated | |
ZXDA | |
ZXDA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7789 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P98168 | |
ZXDA | |
ZXDA Fusion Protein Ag23411 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |