missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AAGAB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27319-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The predicted MW of this protein is 35 kDa. 27319-1-AP recognizes a 37-40 kDa band due to phosphorylation.
The protein encoded by this gene interacts with the gamma-adaptin and alpha-adaptin subunits of complexes involved in clathrin-coated vesicle trafficking. Mutations in this gene are associated with type I punctate palmoplantar keratoderma. Alternatively spliced transcript variants have been found for this gene.Spécification
AAGAB | |
Polyclonal | |
Unconjugated | |
AAGAB | |
AAGAB, FLJ11506 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
66939, 79719 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6PD74, Q8R2R3 | |
Aagab | |
AAGAB Fusion Protein Ag26340 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |