missing translation for 'onlineSavingsMsg'
Learn More

ACK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 14304-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16832884

  • 121.38€ / 20µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Tyrosine kinase. that after binding to CDC42. inhibits both its intrinsic and stimulated GTPase activity. This protein is a tyrosine kinase that binds Cdc42Hs in its GTP-bound form and inhibits both the intrinsic and GTPase-activating protein (GAP).
Spécification

Spécification

ACK1
Polyclonal
Unconjugated
TNK2
ACK, ACK 1, ACK1, Activated CDC42 kinase 1, p21cdc42Hs, TNK2
Rabbit
Antigen Affinity Chromatography
RUO
10188, 303882, 51789
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O54967, Q07912, Q5U2X5
Tnk2
ACK1 Fusion Protein Ag5419
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions