missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADHFE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14674-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The ADHFE1 gene encodes hydroxyacid-oxoacid transhydrogenase (EC 1.1.99.24), which is responsible for the oxidation of 4-hydroxybutyrate in mammalian tissues (Kardon et al., 2006 ).Spécification
ADHFE1 | |
Polyclonal | |
Unconjugated | |
ADHFE1 | |
ADH8, ADHFE1, HMFT2263, HOT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
137872, 362474, 76187 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.65 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q4QQW3, Q8IWW8, Q8R0N6 | |
Adhfe1 | |
ADHFE1 Fusion Protein Ag6382 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |