missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH1L2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21391-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of both the aldehyde dehydrogenase superfamily and the formyl transferase superfamily. This member is the mitochondrial form of 10-formyltetrahydrofolate dehydrogenase (FDH), which converts 10-formyltetrahydrofolate to tetrahydrofolate and CO2 in an NADP(+)-dependent reaction, and plays an essential role in the distribution of one-carbon groups between the cytosolic and mitochondrial compartments of the cell. Alternatively spliced transcript variants have been found for this gene.Spécification
ALDH1L2 | |
Polyclonal | |
Unconjugated | |
ALDH1L2 | |
ALDH1L2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
160428, 216188 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3SY69, Q8K009 | |
ALDH1L2 | |
ALDH1L2 Fusion Protein Ag16046 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |