Learn More
Amylase alpha Mouse anti-Human, Porcine, Rat, Clone: 2F1G8, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66133-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody can recognize amylase alpha.
Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch and glycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas. It is an amylase isoenzyme produced by the salivary gland.Spécification
Amylase alpha | |
Monoclonal | |
0.79 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04746 | |
AMY2A, Amy2a3 | |
Amylase, alpha Fusion Protein Ag8819 | |
20 μL | |
Primary | |
Porcine, Human, Rat | |
Antibody | |
IgG2b |
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
2F1G8 | |
Unconjugated | |
AMY2A | |
AMY2A, Amylase, alpha, Pancreatic alpha amylase | |
Mouse | |
Protein A | |
RUO | |
100521789, 279, 497039 | |
-20°C | |
Liquid |