missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24333-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Recruits CKI-epsilon to the beta-catenin degradation complex that consists of AXN1 or AXN2 and GSK3-beta and allows efficient phosphorylation of beta-catenin, thereby inhibiting beta-catenin/Tcf signals.Spécification
ANKRD6 | |
Polyclonal | |
Unconjugated | |
ANKRD6 | |
ANKRD6, ankyrin repeat domain 6, DIVERSIN, KIAA0957 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22881 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y2G4 | |
ANKRD6 | |
ANKRD6 Fusion Protein Ag19328 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |