missing translation for 'onlineSavingsMsg'
Learn More

CD14 Mouse anti-Human, PE-CF594, Clone: MφP9, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 562335

Code nomenclature Nacres: NA.41

Informations supplémentaires : Poids : 0.00720kg

 Afficher plus de versions de ce produit

Code produit. 15871829

  • 760.00€ / 1 pièce
Expédition estimée: 01-07-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The MφP9 monoclonal antibody specifically binds to human CD14, a 53-55 kDa glycosylphosphatidylinositol (GPI)-anchored and single chain glycoprotein expressed at high levels on monocytes. Additionally, this CD14-specific antibody reacts with interfollicular macrophages, reticular dendritic cells and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB. The purified antibody is suitable for staining acetone-fixed, frozen tissue sections.

This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).

Spécification

Spécification

CD14
Monoclonal
PE-CF594
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human monocytes
100 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
MφP9
LPS receptor; LPS-R; Myeloid cell-specific leucine-rich glycoprotein
Mouse
Affinity Purified
RUO
Human
IgG2b κ
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

For Research Use Only.