Learn More
CD14 Mouse anti-Human, PE-CF594, Clone: MφP9, BD
Mouse Monoclonal Antibody
291.00€ - 760.00€
Spécification
Antigène | CD14 |
---|---|
Clone | MφP9 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugué | PE-CF594 |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15861829
|
BD Biosciences
562334 |
25 Tests |
291.00€
1 pièce |
Expédition estimée: 17-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15871829
|
BD Biosciences
562335 |
100 Tests |
760.00€
1 pièce |
Expédition estimée: 10-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
The MφP9 monoclonal antibody specifically binds to human CD14, a 53-55 kDa glycosylphosphatidylinositol (GPI)-anchored and single chain glycoprotein expressed at high levels on monocytes. Additionally, this CD14-specific antibody reacts with interfollicular macrophages, reticular dendritic cells and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB. The purified antibody is suitable for staining acetone-fixed, frozen tissue sections.
This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).
Spécification
CD14 | |
Flow Cytometry | |
PE-CF594 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG2b κ | |
Affinity Purified | |
LPS receptor; LPS-R; Myeloid cell-specific leucine-rich glycoprotein |
MφP9 | |
Monoclonal | |
Mouse | |
Human | |
Human monocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.