Learn More
CD28 Mouse anti-Human, PE-Cy5, Clone: CD28.2, BD
Mouse Monoclonal Antibody
Marque:  BD Biosciences 561791
Informations supplémentaires : Poids : 0.02000kg
Code produit. 15861339
  • 240.00€ / 1 pièce
Expédition estimée: 22-04-2024
Description
The CD28.2 monoclonal antibody specifically binds to CD28, a 44 kDa homodimeric transmembrane glycoprotein present on most mature T cells, thymocytes and plasma cells. CD28 is a costimulatory receptor that binds CD80 and CD86 as ligands and plays a very important role in T cell-B cell interactions. It has been suggested that CD28 initiates and regulates a separate and distinct signal transduction pathway from those stimulated by the TCR complex. Additionally, it has been reported that CD28 antibody clones vary in their ability to stimulate T cells to produce IL-2 and increase intracellular Ca2+ concentration. This finding suggests the existence of functionally distinct subregions on the CD28 molecule. CD28.2 has been demonstrated to bind to the same molecule as clone L293, another CD28 mAb, and has been reported to induce Ca2+ influx in Jurkat T cells.
Spécification
Spécification
CD28
Monoclonal
PE-Cyanine5
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human CD28 Transfected Cell Line
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
CD28.2
CD28 antigen; T44; Tp44; TP44
Mouse
Affinity Purified
RUO
Human
IgG1 κ
Promotions
For Research Use Only.