Learn More
CD41 Rat anti-Mouse, PE, Clone: MWREG30, BD
Rat Monoclonal Antibody
Marque: BD Biosciences 561850
Informations supplémentaires : Poids : 0.01000kg
Description
The MWReg30 antibody reacts with glycoprotein (gp) IIb, also called integrin αIIb chain (CD41), that associates with integrin β3 chain (gpIIIa or CD61) to form the gpIIb/IIIa (CD41/CD61) complex. CD41/CD61 is expressed on platelets, megakaryocytes, and early hematopoietic progenitors. The integrin complex binds to fibrinogen, fibronectin, vitronectin, von Willebrand factor, and thrombospondin. It is important for platelet adhesion and aggregation, and it may play a role in osteolytic tumor metastasis.
Spécification
CD41 | |
Monoclonal | |
0.2mg/mL | |
Integrin α IIb chain | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
MWREG30 | |
PE | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Platelets | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.