Learn More
TCR β Chain Hamster anti-Mouse, APC, Clone: H57-597, BD
Hamster Monoclonal Antibody
Marque: BD Biosciences 560656
Informations supplémentaires : Poids : 0.01000kg
Description
The H57-597 antibody reacts with a common epitope of the β chain of the T-cell Receptor (TCR) complex on αβ TCR-expressing thymocytes and peripheral T lymphocytes and NK1.1+ thymocytes and NK-T cells of all mouse strains tested. It does not react with γδ TCR-bearing T cells. In the fetal and adult thymus, the TCR β-chain may form homodimers or pair with the pre-TCR α-chain on the surface of immature thymocytes before expression of the TCR α-chain. Plate-bound or soluble H57-597 antibody activates αβ TCR-bearing T cells, and plate-bound mAb can induce apoptotic death.
Flow Cytometry
Spécification
TCR β Chain | |
Monoclonal | |
0.2mg/mL | |
Tcrb; TCRbeta; TCRβ, T cell receptor beta Chain | |
Hamster | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
H57-597 | |
APC | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
TCR affinity-purified from mouse T-cell hybridoma DO-11.10 | |
50 μg | |
Immunology | |
Mouse | |
IgG2 |
For Research Use Only.