missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AP3D1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16454-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
AP3D1 is a subunit of the AP3 adaptor-like complex, which is not associated with clathrin. The AP3D1 subunit is implicated in intracellular biogenesis and trafficking of pigment granules and possibly platelet dense granules and neurotransmitter vesicles.Spécification
AP3D1 | |
Polyclonal | |
Unconjugated | |
AP3D1 | |
ADTD, AP 3 complex subunit delta, AP 3 complex subunit delta 1, AP3D1, Delta adaptin, hBLVR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11776, 314633, 8943 | |
-20°C | |
Liquid |
Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14617, O54774 | |
Ap3d1 | |
AP3D1 Fusion Protein Ag9390 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |