missing translation for 'onlineSavingsMsg'
Learn More

APOBEC4 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 17166-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837004

  • 121.38€ / 20µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a member of the AID/APOBEC family of polynucleotide (deoxy)cytidine deaminases, which convert cytidine to uridine. Other AID/APOBEC family members are involved in mRNA editing, somatic hypermutation and recombination of immunoglobulin genes, and innate immunity to retroviral infection.
Spécification

Spécification

APOBEC4
Polyclonal
Unconjugated
APOBEC4
APOBEC4, C1orf169
Rabbit
Antigen Affinity Chromatography
RUO
403314
-20°C
Liquid
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin)
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8WW27
APOBEC4
APOBEC4 Fusion Protein Ag10633
20 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions