missing translation for 'onlineSavingsMsg'
Learn More

ARCN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 23843-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16842475

  • 140.00€ / 20µL
Expédition estimée: 27-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene maps in a region, which include the mixed lineage leukemia and Friend leukemia virus integration 1 genes, where multiple disease-associated chromosome translocations occur. It is an intracellular protein. Archain sequences are well conserved among eukaryotes and this protein may play a fundamental role in eukaryotic cell biology. It has similarities to heat shock proteins and clathrin-associated proteins, and may be involved in vesicle structure or trafficking.
Spécification

Spécification

ARCN1
Polyclonal
Unconjugated
ARCN1
Archain, archain 1, ARCN1, Coatomer subunit delta, COPD, Delta coat protein, Delta COP
Rabbit
Antigen Affinity Chromatography
RUO
213827, 372
-20°C
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P48444, Q5XJY5
ARCN1
ARCN1 Fusion Protein Ag20836
20 μL
Primary
Human, Mouse
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions