missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGEF4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 55213-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody is specific to ARHGEF4.
Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form complex with G proteins and stimulate Rho-dependent signals. This protein is similar to rat collybistin protein. Alternative splicing of this gene generates two transcript variants which encode different isoforms. Also there is possibility for the usage of multiple polyadenylation sites for this gene.Spécification
ARHGEF4 | |
Polyclonal | |
Unconjugated | |
ARHGEF4 | |
ARHGEF4, ASEF, ASEF1, GEF4, KIAA1112, STM6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
226970, 50649 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7TNR9, Q9NR80 | |
ARHGEF4 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |