missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARPC5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16717-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p16 subunit, has yet to be determined.Spécification
ARPC5 | |
Polyclonal | |
Unconjugated | |
ARPC5 | |
ARC16, ARPC5, p16 Arc | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10092, 360854, 67771 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15511, Q4KLF8, Q9CPW4 | |
Arpc5 | |
ARPC5 Fusion Protein Ag10144 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |