missing translation for 'onlineSavingsMsg'
Learn More

Arrestin C Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 11100-2-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837953

  • 325.38€ / 150µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Arrestin-C, also known as retinal cone Arrestin-3, X-Arrestin or cArr, is a member of the Arrestin family of proteins. It is predomitly found in the retina and pineal gland and localizes to the inner and outer segments of red-, green- and blue-cone photoreceptors and the inner plexiform regions. Two Arrestin-C isoforms exist due to alternative splicing. Isoform 1 is the mature full length protein and isoform 2 is truncated, ending with an arginine for amino acid residue 359. Arrestin-C expression is stimulated by retinoic acid. It may play a role in retina-specific signal transduction and bind to photoactivated-phosphorylated red/green opsins. In addition, Arrestin-C forms homodimers and oligomers with beta-Arrestins and may regulate beta-Arrestin mediated signaling.
Spécification

Spécification

Arrestin C
Polyclonal
Unconjugated
ARR3
ARR3, Arrestin C, ARRX, C arrestin, CAR, cArr, Cone arrestin, Retinal cone arrestin 3, X arrestin
Rabbit
Antigen Affinity Chromatography
RUO
170735, 407
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P36575, Q9EQP6
ARR3
Arrestin C Fusion Protein Ag1580
150 μL
Primary
Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions