missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATXN3L Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25099-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
ATXN3L also named as ataxin 3 like or MJDL is a 355 amino acid protein, which contains one Josephin domain and two UIM repeats. ATXN3L localizes in nucleus and may cleave poly ubiquitin chain.Spécification
ATXN3L | |
Polyclonal | |
Unconjugated | |
ATXN3L | |
ataxin 3 like, ATX3L, ATXN3L, MJDL, Putative ataxin 3 like protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
92552 | |
-20°C | |
Liquid |
Western Blot | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H3M9 | |
ATXN3L | |
ATXN3L Fusion Protein Ag18740 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |