missing translation for 'onlineSavingsMsg'
Learn More

BACE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 16321-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16835914

  • 399.00€ / 150µL
Expédition estimée: 15-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Cerebral deposition of amyloid beta peptide is an early and critical feature of Alzheimer′s disease and a frequent complication of Down syndrome. Amyloid beta peptide is generated by proteolytic cleavage of amyloid precursor protein by 2 proteases, one of which is the protein encoded by this gene. This gene localizes to the ′Down critical region′ of chromosome 21. The encoded protein, a member of the peptidase A1 protein family, is a type I integral membrane glycoprotein and aspartic protease. Three transcript variants encoding different isoforms have been described for this gene.
Spécification

Spécification

BACE2
Polyclonal
Unconjugated
BACE2
AEPLC, ALP56, Asp 1, ASP1, ASP21, Aspartic like protease 56 kDa, Aspartyl protease 1, BACE2, BAE2, Beta secretase 2, CDA13, CEAP1, Down region aspartic protease, DRAP, Memapsin 1, Theta secretase, UNQ418/PRO852
Rabbit
Antigen Affinity Chromatography
RUO
25825, 288227, 56175
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6IE75, Q9JL18, Q9Y5Z0
Bace2
BACE2 Fusion Protein Ag9404
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions