Learn More
BANP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 16393-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16836004
  • 325.38€ / 150µL
Expédition estimée: 28-09-2022
Description
This gene encodes a protein that binds to matrix attachment regions. The protein forms a complex with p53 and negatively regulates p53 transcription, and functions as a tumor suppressor and cell cycle regulator. Multiple transcript variants encoding different isoforms have been found for this gene.
Spécification
Spécification
BANP
Polyclonal
Unconjugated
BANP
BANP, BEND1, Protein BANP, SMAR1, SMARBP1
Rabbit
Antigen Affinity Chromatography
RUO
292064, 53325, 54971
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8N9N5, Q8VBU8
Banp
BANP Fusion Protein Ag9484
150 μL
Primary
Rat, Mouse, Human