missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17254-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
BBX gene ontology annotations related to this gene include DNA binding.Spécification
BBX | |
Polyclonal | |
Unconjugated | |
BBX | |
BBX, Bobby sox homolog, bobby sox homolog (Drosophila), HBP2, HMG box containing protein 2, HSPC339, MDS001 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
303970, 56987, 70508 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8VBW5, Q8WY36 | |
Bbx | |
BBX Fusion Protein Ag10691 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |