missing translation for 'onlineSavingsMsg'
Learn More

BTN1A1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 26687-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16835085

  • 399.00€ / 150µL
Expédition estimée: 30-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The calculated molecular weight of BTN1A1 is a 59 kDa, but the modified BTN1A1 protein is about 65-75 kDa.

BTN1A1 (Butyrophilin) is the major protein associated with fat droplets in the milk. It is a member of the immunoglobulin superfamily. It may have a cell surface receptor function. The human butyrophilin gene is localized in the major histocompatibility complex (MHC) class I region of 6p and may have arisen relatively recently in evolution by the shuffling of exons between 2 ancestral gene families.
Spécification

Spécification

BTN1A1
Polyclonal
Unconjugated
BTN1A1
BT, BTN, BTN1A1
Rabbit
Antigen Affinity Chromatography
RUO
696
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q13410
BTN1A1
BTN1A1 Fusion Protein Ag24613
150 μL
Primary
Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions