missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTN1A1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26687-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The calculated molecular weight of BTN1A1 is a 59 kDa, but the modified BTN1A1 protein is about 65-75 kDa.
BTN1A1 (Butyrophilin) is the major protein associated with fat droplets in the milk. It is a member of the immunoglobulin superfamily. It may have a cell surface receptor function. The human butyrophilin gene is localized in the major histocompatibility complex (MHC) class I region of 6p and may have arisen relatively recently in evolution by the shuffling of exons between 2 ancestral gene families.Spécification
BTN1A1 | |
Polyclonal | |
Unconjugated | |
BTN1A1 | |
BT, BTN, BTN1A1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
696 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13410 | |
BTN1A1 | |
BTN1A1 Fusion Protein Ag24613 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |