Learn More
c-Myb Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17800-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Transcriptional activator; DNA-binding protein that specifically recognize the sequence 5′-YAAC[GT]G-3′. Plays an important role in the control of proliferation and differentiation of hematopoietic progenitor cells.Spécification
c-Myb | |
Polyclonal | |
Unconjugated | |
MYB | |
c myb_CDS, Cmyb, c-Myb, efg, MYB, Proto oncogene c Myb, Transcriptional activator Myb | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17863, 4602 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P06876, P10242 | |
Myb | |
c-Myb Fusion Protein Ag12021 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |