missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C10orf27 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24424-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
May play a role in spermatid differentiation. Modulates thymic stromal cell proliferation and thymus function.Spécification
C10orf27 | |
Polyclonal | |
Unconjugated | |
TBATA | |
C10orf27, Protein SPATIAL, spatial, TBATA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
219793 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96M53 | |
TBATA | |
C10orf27 Fusion Protein Ag19625 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |