missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C13orf27/TEX30 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25408-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
C13orf27 is a protein coding gene.Spécification
C13orf27/TEX30 | |
Polyclonal | |
Unconjugated | |
TEX30 | |
C13orf27, TEX30 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
75623, 93081 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3TUU5, Q5JUR7 | |
Tex30 | |
C13orf27/TEX30 Fusion Protein Ag22096 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |