missing translation for 'onlineSavingsMsg'
Learn More

C19orf43 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 19420-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16888974

  • 349.00€ / 150µL
Expédition estimée: 16-12-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The antibody was raised from the C19orf43 protein. A band at ∽28kd was detected in Western Blot with human placenta, 293 cell, and A431 cell lysates.

C19orf43, also named as MGC2803 and fSAP18, is a protein coding gene. Exoribonuclease that is part of the telomerase RNA 3′ end processing complex and which has the ability to all four unpaired RNA nucleotides from 5′ end or 3′ end with higher efficiency for purine bases.
Spécification

Spécification

C19orf43
Polyclonal
Unconjugated
C19orf43
C19orf43, fSAP18
Rabbit
Antigen Affinity Chromatography
RUO
288920, 68544, 79002
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BQ61
C19orf43, Trir
C19orf43 Fusion Protein Ag13747
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions