missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C3orf34 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26036-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene localizes to centrosomes and primary cilia and co-localizes with a marker for the mother centriole. This gene resides in a region of human chromosome 3 that is linked to morbid obesity. A homozygous knockout of the orthologous gene in mouse resulted in mice with morbid obesity, hyperphagy, glucose intolerance, and insulin resistance. Mutations in this gene cause morbid obesity and spermatogenic failure (MOSPGF). This gene has a pseudogene on human chromosome 2.Spécification
C3orf34 | |
Polyclonal | |
Unconjugated | |
CEP19 | |
C3orf34, CEP19, UPF0709 protein C3orf34 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
66994, 84984 | |
-20°C | |
Liquid |
Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96LK0, Q9CQA8 | |
CEP19 | |
C3orf34 Fusion Protein Ag22035 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |