Learn More
CABC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15528-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a mitochondrial protein similar to yeast ABC1, which functions in an electron-transferring membrane protein complex in the respiratory chain. It is not related to the family of ABC transporter proteins. Expression of this gene is induced by the tumor suppressor p53 and in response to DNA damage, and inhibiting its expression partially suppresses p53-induced apoptosis. Alternatively spliced transcript variants have been found; however, their full-length nature has not been determined.Spécification
CABC1 | |
Polyclonal | |
Unconjugated | |
COQ8A | |
ADCK3, ARCA2, CABC1, Chaperone ABC1 like, COQ8, SCAR9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
360887, 56997, 67426 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5BJQ0, Q60936, Q8NI60 | |
Coq8a | |
CABC1 Fusion Protein Ag7612 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |