missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catenin, beta Mouse, Clone: 5H10, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 1499-MSM4-P1
Informations supplémentaires : Poids : 0.01000kg
Spécification
Catenin, beta | |
Cell surface and cytoplasmic | |
5H10 | |
Unconjugated | |
NP_001895 | |
Mouse | |
92kDa | |
Protein A/G Purified | |
RUO | |
1499 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
CTNNB1 | |
Fusion protein consisting of the maltose binding protein fused to a 100 amino acid segment of the C-terminus of chicken beta-Catenin | |
HeLa or MCF-7 cells. Breast carcinoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |