missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC28B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11530-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The product of this gene localizes to centrosomes and basal bodies. It interacts and colocalizes with several proteins associated with Bardet-Biedl syndrome (BBS).Spécification
CCDC28B | |
Polyclonal | |
Unconjugated | |
CCDC28B | |
CCDC28B, RP4 622L5.5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
66264, 682445, 79140 | |
-20°C | |
Liquid |
Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8CEG5, Q9BUN5 | |
Ccdc28b | |
CCDC28B Fusion Protein Ag2105 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |