missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR7 (CD197) Mouse anti-Human , BV650, Clone: 2-L1-A, BD Biosciences™
Mouse Monoclonal Antibody
Marque: BD Biosciences 566757
Informations supplémentaires : Poids : 0.00020kg
Description
CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow CytometrySpécification
CCR7 (CD197) | |
Monoclonal | |
Brilliant Violet 650 | |
BLR2; CC chemokine receptor 7; CMKBR7; EBI1; EVI1; Epstein-Barr virus induced gene 1; MIP-3 beta receptor | |
Human CCR7 Transfected Cell Line | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
2-L1-A | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV650 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV650 were removed. |