Learn More
CD100 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27516-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The CD100 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD100. This antibody recognizes human antigen. The CD100 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
CD100 | |
Polyclonal | |
Unconjugated | |
150 μL | |
10507 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
267 μg/mL | |
Rabbit | |
Primary | |
Human |