Learn More
CD100 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 27516-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16835675
  • 399.00€ / 150µL
Expédition estimée: 15-04-2024
Description
The CD100 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD100. This antibody recognizes human antigen. The CD100 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
Spécification
CD100
Polyclonal
Unconjugated
150 μL
10507
IgG
Western Blot, Immunohistochemistry, ELISA
267 μg/mL